5-Fluororotsäure

Lieferant: Serva

Synonyme: 5-FOA

21650.02
SERA21650.02EA 0 EUR
SERA21650.02
5-Fluororotsäure
5-Fluororotsäure
Formel: C₅H₃FN₂O₄
Molecular Weight: 174.09 g/mol
MDL: MFCD00042526
CAS-Nummer: 703-95-7
EINECS: 211-876-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR