D(+)-Galactosamin Hydrochlorid
Lieferant: Serva
Synonyme:
(2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
22009.03
SERA22009.03EA
0
EUR
SERA22009.03
D(+)-Galactosamin Hydrochlorid
D(+)-Galactosamin Hydrochlorid
Formel:
C₆H₁₄ClNO₅ Molecular Weight: 215,64 g/mol |
MDL:
MFCD00135830 CAS-Nummer: 1772-03-8 EINECS: 217-198-1 |