Base materials
Lieferant: Serva
24540.02 24540.03 48600.02 48600.04 48602.05 48610.05 48605.01 48618.01 48618.02 48619.01 48619.02 48622.01 48622.02
SERA24540.02EA 93.8 EUR
SERA24540.02 SERA24540.03 SERA48600.02 SERA48600.04 SERA48602.05 SERA48610.05 SERA48605.01 SERA48618.01 SERA48618.02 SERA48619.01 SERA48619.02 SERA48622.01 SERA48622.02
Base materials
Medien für die Mikrobiologie Basismaterialien für die Mikrobiologie
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation