L(+)-Histidin Hydrochlorid Monohydrat
Lieferant: Serva
Synonyme:
His HCl Monohydrat, (S)-(+)-Histidin Hydrochlorid Monohydrat, (2S)-(+)-Histidin Hydrochlorid Monohydrat, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, H-His-OH.HCl.H2O
24842.02
SERA24842.02EA
109
EUR
SERA24842.02
L(+)-Histidin Hydrochlorid Monohydrat
L(+)-Histidin Hydrochlorid Monohydrat
Formel:
C₆H₉N₃O₂·HCl·H₂O Molecular Weight: 209,63 g/mol Schmelzpunkt: 259 °C Dichte: 1,485 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151027 CAS-Nummer: 5934-29-2 EINECS: 211-438-9 Merck Index: 13,04741 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...