Hydroxylammoniumchlorid
Lieferant: Serva
|
Achtung
|
Synonyme:
Hydroxylamin-Hydrochlorid, Chlorhydrathydroxylamin, Hydroxylamin Hydrochlorid, Hydroxylamin HCl
25513.02
SERA25513.02EA
0
EUR
SERA25513.02
Hydroxylammoniumchlorid
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Molecular Weight: 69,49 g/mol Siedepunkt: 306 °C (1013 hPa) Schmelzpunkt: 151 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III Merck Index: 13,04853 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...