Ionomycin Calciumsalz

Lieferant: Serva

26379.01
SERA26379.01EA 0 EUR
SERA26379.01
Ionomycin Calciumsalz
Ionomycin calcium salt
Formel: C₄₁H₇₀CaO₉
Molecular Weight: 747.1 g/mol
MDL: MFCD00151183
CAS-Nummer: 56092-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR