3-Isobutyl-1-methylxanthin

Lieferant: Serva
Achtung

Synonyme: 3-Isobutyl-1-methyl-2,6(1H,3H)-purindion, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-1-methyl-3-isobutylpurin

26445.02
SERA26445.02EA 141 EUR
SERA26445.02
3-Isobutyl-1-methylxanthin
3-Isobutyl-1-methylxanthin
Formel: C₁₀H₁₄N₄O₂
Molecular Weight: 222.25 g/mol
Schmelzpunkt: 200…201 °C
MDL: MFCD00005584
CAS-Nummer: 28822-58-4
EINECS: 249-259-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR