Kongorot
Lieferant: Serva
|
Gefahr
|
Synonyme:
Direktrot 28, Direct Red 28, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)
27215.01
SERA27215.01EA
57.4
EUR
SERA27215.01
Kongorot
Kongorot
Formel:
C₃₂H₂₂N₆Na₂O₆S₂ Molecular Weight: 696,67 g/mol Schmelzpunkt: > 360 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004028 CAS-Nummer: 573-58-0 EINECS: 209-358-4 Merck Index: 12,02562 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...