Lanthan(III)nitrat

Lieferant: Serva
Gefahr

27507.01
SERA27507.01EA 0 EUR
SERA27507.01
Lanthan(III)nitrat
Lanthan(III)nitrat
Formel: LaN₃O₉
Molecular Weight: 324,92 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011067
CAS-Nummer: 10099-59-9
EINECS: 233-238-0
UN: 1477
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR