L(+)-Leucin

Lieferant: Serva

Synonyme: H-Leu-OH, (S)-(+)-α-Amino-γ-methylvaleriansäure, L-Leucin, (S)-(+)-Leucin, (S)-(+)-2-Amino-4-methylpentansäure

27690.02 27690.03
SERA27690.02EA 0 EUR
SERA27690.02 SERA27690.03
L(+)-Leucin
L(+)-Leucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
Schmelzpunkt: 300
Dichte: 1.165 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002617
CAS-Nummer: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR