D(-)-Luciferin

Lieferant: Serva

28073.01 28073.02 28073.03
SERA28073.01EA 0 EUR
SERA28073.01 SERA28073.02 SERA28073.03
D(-)-Luciferin
D(-)-Luciferin
Formel: C₁₁H₈N₂O₃S₂
MDL: MFCD00042929
CAS-Nummer: 2591-17-5
EINECS: 219-981-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR