Pig Malate dehydrogenase (from Heart)
Lieferant: Serva
28338.02
SERA28338.02EA 0 EUR
SERA28338.02
Pig Malate dehydrogenase (from Heart)
Enzyme
A highly purified suspension in 80% saturated ammonium sulphate. Suitable for GOT determination in serum.