Pig Malate dehydrogenase (from Heart)
Lieferant: Serva
28345.01
SERA28345.01EA 0 EUR
SERA28345.01
Pig Malate dehydrogenase (from Heart)
Enzyme
An ammonium sulphate free solution in 50% glycerol. 5 mg correspond to approx. 0,5 ml.