Naphthol AS-E-phosphat
Lieferant: Serva
Synonyme:
N-(4-Chlorphenyl)-3-(phosphonooxy)naphthalin-2-carboxamid
29998.02
SERA29998.02EA
0
EUR
SERA29998.02
Naphthol AS-E-phosphat
Naphthol AS-E-phosphat (KG-501)
Formel:
C₁₇H₁₃ClNO₅P Molecular Weight: 377,72 g/mol |
CAS-Nummer:
18228-17-6 EINECS: 242-106-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...