Neufuchsin
Lieferant: Serva
|
Gefahr
|
Synonyme:
4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-yliden)methyl]-o-toluidin Hydrochlorid, 4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-yliden)methyl]-o-toluidiniumchlorid, 4-(Di(4-amino-3-methylphenyl)methylen)-2-methyl-2,5-cyclohexadien-1-yliden)ammoniumchlorid, 4,4',4''-Triamino-3,3',3''-trimethyltrityliumchlorid
30293.01
SERA30293.01EA
0
EUR
SERA30293.01
Neufuchsin
Neufuchsin
Formel:
C₂₂H₂₄ClN₃ Molecular Weight: 365,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012567 CAS-Nummer: 3248-91-7 EINECS: 221-831-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...