β-Nicotinamid-Adenin-Dinucleotid-Phosphat, (β-NADP)

Lieferant: Serva

Synonyme: Nicotinamid-Adenin-Dinucleotid-Phosphat, oxidiert, Nadidphosphat, Nicotinamid-Adenin-Dinucleotid-Phosphat, TPN, β-Nicotinamid-Adenin-Dinucleotid-Phosphat

30313.01 30313.02
SERA30313.01EA 0 EUR
SERA30313.01 SERA30313.02
β-Nicotinamid-Adenin-Dinucleotid-Phosphat, (β-NADP)
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat, oxidierte Form (NADP)
Formel: C₂₁H₂₈N₇O₁₇P₃
Molecular Weight: 743.41 g/mol
MDL: MFCD00067537
CAS-Nummer: 53-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR