4-(4-Nitrobenzyl)pyridin

Lieferant: Serva

Synonyme: 4-(p-Nitrobenzyl)Pyridine

30548.02
SERA30548.02EA 0 EUR
SERA30548.02
4-(4-Nitrobenzyl)pyridin
4-(4-Nitrobenzyl)pyridin
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006445
CAS-Nummer: 1083-48-3
EINECS: 214-108-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR