AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)

Lieferant: Serva
Achtung

Synonyme: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

31682.01 31682.02
SERA31682.01EA 99.9 EUR
SERA31682.01 SERA31682.02
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Formel: C₈H₁₁ClFNO₂S
Molecular Weight: 239.69 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00132962
CAS-Nummer: 30827-99-7
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR