1-Pentansulfonsäure Natriumsalz
Lieferant: Serva
Synonyme:
Natriumpentylsulfonat
31812.02
SERA31812.02EA
0
EUR
SERA31812.02
1-Pentansulfonsäure Natriumsalz
1-Pentanesulphonic acid sodium salt
Formel:
C₅H₁₁O₃SNa Molecular Weight: 174,2 g/mol Schmelzpunkt: > 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007541 CAS-Nummer: 22767-49-3 EINECS: 245-208-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...