Progesteron

Lieferant: Serva

33530.03
SERA33530.03EA 0 EUR
SERA33530.03
Progesteron
Progesteron
Formel: C₂₁H₃₀O₂
MDL: MFCD00003658
CAS-Nummer: 57-83-0
EINECS: 200-350-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR