Silicone DC 710 fluid
Lieferant: Serva
35149.04
SERA35149.04EA 497 EUR
SERA35149.04
Silicone DC 710 fluid
Flüssigkeiten für Umwälzbäder
  • Retains its properties after continuous use at temperatures up to 260°C in closed systems
  • Shows very low volatility even at elevated temperatures
  • Highly resistant to oxidation and gumming