Taurin

Lieferant: Serva

Synonyme: 2-Aminoethanesulphonic acid

35770.03 35770.04
SERA35770.03EA 0 EUR
SERA35770.03 SERA35770.04
Taurin
Taurin
Formel: C₂H₇NO₃S
Molecular Weight: 125.15 g/mol
Schmelzpunkt: 300
Lagertemperatur: Raumtemperatur
MDL: MFCD00008197
CAS-Nummer: 107-35-7
EINECS: 203-483-8
Merck Index: 13,09163

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR