Tetranitroblau-Tetrazoliumchlorid

Lieferant: Serva

Synonyme: TNBT

35935.02 35935.03
SERA35935.02EA 0 EUR
SERA35935.02 SERA35935.03
Tetranitroblau-Tetrazoliumchlorid
Tetranitroblau-Tetrazoliumchlorid
Formel: C₄₀H₂₈Cl₂N₁₂O₁₀
Molecular Weight: 907.64 g/mol
Schmelzpunkt: 165
MDL: MFCD00036338
CAS-Nummer: 1184-43-6
EINECS: 214-665-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR