Thiamin Hydrochlorid

Lieferant: Serva

36020.02 36020-0100
SERA36020.02EA 85.6 EUR
SERA36020.02 SERA36020-0100
Thiamin Hydrochlorid
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Schmelzpunkt: 252 °C
Dichte: 1,401 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR