L-Threonin

Lieferant: Serva

Synonyme: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

36382.02 36382.03
SERA36382.02EA 0 EUR
SERA36382.02 SERA36382.03
L-Threonin
L-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064270
CAS-Nummer: 72-19-5
EINECS: 200-774-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR