Triethanolamin Hydrochlorid

Lieferant: Serva

36885.02
SERA36885.02EA 0 EUR
SERA36885.02
Triethanolamin Hydrochlorid
Triethanolamin Hydrochlorid
Formel: C₆H₁₆ClNO₃
Molecular Weight: 185,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012596
CAS-Nummer: 637-39-8
EINECS: 211-284-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR