2,4,6-Tri-(dimethylaminomethyl)phenol
Lieferant: Serva
Synonyme:
Epon® DMP-30 Substitut, DMP-30®, Epoxy-Einbettungsmittel, Beschleuniger
36975.01
SERA36975.01EA
39.6
EUR
SERA36975.01
2,4,6-Tri-(dimethylaminomethyl)phenol
2,4,6-Tri-(dimethylaminomethyl)phenol
Formel:
C₁₅H₂₇N₃O Molecular Weight: 265,4 g/mol Siedepunkt: 130…135 °C (1 mmHg) Dichte: 0,974 g/cm³ (15 °C) |
MDL:
MFCD00008330 CAS-Nummer: 90-72-2 EINECS: 202-013-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...