Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)

Lieferant: Serva
Achtung

Synonyme: Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol

37180.02 37180.03 37180.04 37180.05
SERA37180.02EA 37.2 EUR
SERA37180.02 SERA37180.03 SERA37180.04 SERA37180.05
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel: NH₂C(CH₂OH)₃
Molecular Weight: 121.14 g/mol
Siedepunkt: 357
Schmelzpunkt: 169…172 °C
Dichte: 1.328 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004679
CAS-Nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR