TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)

Lieferant: Serva

Synonyme: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

37194.02 37194.03
SERA37194.02EA 0 EUR
SERA37194.02 SERA37194.03
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)
Formel: C₇H₁₇NO₆S
Molecular Weight: 243.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007538
CAS-Nummer: 29915-38-6
EINECS: 249-954-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR