Tricin
Lieferant: Serva
Synonyme:
N-(Tris(hydroxymethyl)methyl)glycin
37195.01
37195.03
SERA37195.01EA
0
EUR
SERA37195.01
SERA37195.03
Tricin
Tricin
Formel:
(HOCH₂)₃CNHCH₂CO₂H Schmelzpunkt: 187 °C Dichte: 1,523 g/cm³ (19.85 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004277 CAS-Nummer: 5704-04-1 EINECS: 227-193-6 Merck Index: 13,09724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...