Vitamin B12 (Cyanocobalamin)

Lieferant: Serva

38310.02
SERA38310.02EA 70.5 EUR
SERA38310.02
Vitamin B12 (Cyanocobalamin)
Vitamin B12 (Cyanocobalamin)
Formel: C₆₃H₈₈CoN₁₄O₁₄P
Molecular Weight: 1355.38 g/mol
Schmelzpunkt: 320
Lagertemperatur: Kühlschrank
MDL: MFCD00151092
CAS-Nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR