Zedernholzöl

Lieferant: Serva
Achtung

Synonyme: Cedar wood oil, Cedar Oil, Cedar Leaf Oil

38565.01
SERA38565.01EA 0 EUR
SERA38565.01
Zedernholzöl
Oil of cedar wood
Dichte: 0,98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00146327
CAS-Nummer: 8000-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR