Chloroform : isoamyl alcohol (24:1) (v/v)

Lieferant: Serva
Achtung

39554.01
SERA39554.01EA 0 EUR
SERA39554.01
Chloroform : isoamyl alcohol (24:1) (v/v)
Chloroform : isoamyl alcohol
Formel: CHCl₃,C₅H₁₂O
Lagertemperatur: Raumtemperatur
UN: 1888
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR