Polyvinylpyrrolidon, M.W. ∼ 360,000 (K90)

Lieferant: Serva

Synonyme: Polyvidon, Poly[1-(2-oxo-1-pyrrolidinyl)ethylen], PVP

39609.01 39609.02
SERA39609.01EA 0 EUR
SERA39609.01 SERA39609.02
Polyvinylpyrrolidon, M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR