Ammoniumchlorid
Lieferant: Serva
|
Achtung
|
Synonyme:
Salmiak
39752.01
SERA39752.01EA
82.1
EUR
SERA39752.01
Ammoniumchlorid
Ammoniumchlorid
Formel:
NH₄Cl Molecular Weight: 53.49 g/mol Schmelzpunkt: 335 Dichte: 1.5256 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011420 CAS-Nummer: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...