Magnesiumchlorid 1 mol/l (2 N) aqueous solution
Lieferant: Serva
39772.01
39772.02
SERA39772.01EA
37.3
EUR
SERA39772.01
SERA39772.02
Magnesiumchlorid 1 mol/l (2 N) aqueous solution
Magnesium chloride
Formel:
MgCl₂ Molecular Weight: 95.21 g/mol Siedepunkt: 1412 Schmelzpunkt: 715 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011106 CAS-Nummer: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...