Tween® 20 (Polysorbat)

Lieferant: Serva

39796.01
SERA39796.01EA 50.3 EUR
SERA39796.01
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)
Formel: C₅₈H₁₁₄O₂₆
Dichte: 1,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
EINECS: 500-018-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR