Precast horizontal gel kits, SERVALYT™ PRECOTES™ Hp Kit
Lieferant: Serva
42805.01
SERA42805.01EA 0 EUR
SERA42805.01
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Hp Kit
Elektrophorese-Gele Fertiggele
For haptoglobin (Hp) analysis by isoelectric focusing.
Lieferumfang: SERVALYT™ PRECOTES™ Hp Kit contains:
5× SERVALYT™ PRECOTES™ Hp gels, 245×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks