Agarose

Lieferant: Serva

11404.02 11404.03 11404.04 11404.05 A11404.04 11404.07
SERA11404.02EA 0 EUR
SERA11404.02 SERA11404.03 SERA11404.04 SERA11404.05 SERVA11404.04 SERA11404.07
Agarose
Agarose
Schmelzpunkt: 60…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081294
CAS-Nummer: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR