Hotplates
Lieferant: SEVERIN ELECTROGERAETE
DK1042
SEVEDK1042EA 142 EUR
SEVEDK1042
Hotplates
Heizgeräte Heizplatten
This hotplate features two heating elements. The exterior is white highly heat-resistant paint.
  • Stepless temperature control and pilot lights