Bag sealer, electronic
Lieferant: SEVERIN ELECTROGERAETE
FS3604
SEVEFS3604EA 87.4 EUR
SEVEFS3604
Bag sealer, electronic
Heißversiegelungsgeräte Beutelverschlussgeräte
For vacuum sealing bags.
  • Air suction function
  • Sealing wire and cut-off wire
  • Viewing window
  • Pilot lights for both vacuum control and sealing function
  • Roll of sealing film (width 28,5 cm) included
  • Cable storage
W×D×H: 355×142×100 mm