Water purification system, Ultra Clear™ RO DI
Lieferant: EVOQUA WATER TECHNOLOGIES
3000-DI
SGWA3000-DIEA 0 EUR
SGWA3000-DI
Water purification system, Ultra Clear™ RO DI
Wasseraufbereitungssysteme Reinstwassersysteme
The Ultra Clear™ RO DI series system delivers permeate water with a salt reduction of approximately 98%.
  • Lower cost alternative to electrodeionisation systems
  • Delivers water with a quality that fulfills the needs of most laboratory requirements
  • Display inlet and product water quality
  • Salt reduction rate will be shown in percentage
The Ultra Clear™ RO DI Series System will polish the RO permeate to a quality of <0,1 S/cm. At a 98% rejection rate, the RO permeate water that originated from a 400 S/cm feed water source would still have a conductivity of about 8 S/cm.
The deionisation module is delivered with the Ultra Clear™ RO DI Series System. These modules are directional and will require replacing at a frequency depending on feed water quality and water consumption. A changing interval of about 3 months is recommended.