Ultrapure water system, LaboStar® Pro
Lieferant: EVOQUA WATER TECHNOLOGIES
W3T324339
SGWAW3T324339EA 4680 EUR
SGWAW3T324339
Ultrapure water system, LaboStar® Pro
Wasseraufbereitungssysteme Reinstwassersysteme
The LaboStar® PRO system is a cost effective means of producing analytical grade water. This extremely compact ultrapure water system can be used on a laboratory bench.
  • Whisper operation mode
  • Pressure reducer included
  • Protective jacket at the end of the sterile filter
  • Ultrapure water circulation right into the dispenser head
  • Quality monitoring after each purification stage
  • Easy to dispense water using the practical dispenser
LaboStar® system is fed with either deionized water, distilled water or permeate from a reverse osmosis unit. The selection of treatment materials in 2 and 4 systems, along with the DI polishing modules, guarantees that the product water meets the highest quality standards. A conductivity meter in the recirculation section continually monitors the purity of the product water.