Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Lieferant: HyClone products (Cytiva)
SH30256.01
SH30258.01
SH30256.02
SH30256.FS
SH30256.LS
SH30258.02
SH30256.01EA
43.4
EUR
SH30256.01
HYCLSH30258.01
HYCLSH30256.02
HYCLSH30256.FS
HYCLSH30256.LS
HYCLSH30258.02
Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Puffer
Gewebekultur-Puffer
Balanced salt solution used for multiple cell culture applications.
- Full traceability and documented origin of all formula ingredients
- Validated and consistent production processes
0,1 µm sterile filtered
Endotoxin Concentration: ≤1.0 EU/mL
Standards: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities