Wasser, HyClone™ HyPure, Molecular Biology Grade Molecular biology grade

Lieferant: HyClone products (Cytiva)

Synonyme: Wasserstoffoxid, Hydrogenoxid

SH30538.01 SH30538.02 SH30538.03 SH30538.FS SH30538.LS
SH30538.01EA 27.6 EUR
SH30538.01 HYCLSH30538.02 HYCLSH30538.03 HYCLSH30538.FS HYCLSH30538.LS
Wasser, HyClone™ HyPure, Molecular Biology Grade Molecular biology grade
Wasser

Nuclease-free HyClone Molecular Biology-Grade Water, containing no DEPC or other carcinogenic chemicals is used for critical applications involving nucleotides such as RNA and DNA.


  • Significantly low levels of endotoxins
  • Certified to have no detectable activity in DNase, RNase, and protease assays
  • Maximum allowable limit of endotoxin is 0.05 EU/mL


HyClone HyPure Molecular Biology Grade Water purification process includes continuous deionization, reverse osmosis, ultraviolet light treatment, distillation, and 0.1 µm filtration.


Standards: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).filtration

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Endotoxin Concentration ≤ 0,05 EU/mL
Sterilisation 0,1 µm sterile filtered
pH 5,0 to 7,0
Storage 2° to 30° C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR