Natriumhydroxid solution_SC 0.1 N (0.1 M), single-use containers, HyClone™ Bioprocessing

Lieferant: HyClone products (Cytiva)

Synonyme: Caustic soda

SH30883.01 SH30883.03 SH30883.04
SH30883.01EA 632 EUR
SH30883.01 SH30883.03 SH30883.04
Natriumhydroxid solution_SC 0.1 N (0.1 M), single-use containers, HyClone™ Bioprocessing
Sodium hydroxide

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.


  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 µm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.


HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Storage 15° to 30°C
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 Months
pH For Information Only
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR