Cell culture medium, HyClone™ ActiPro
Lieferant: HyClone products (Cytiva)
SH31037.01 SH31039.04 SH31037.08 SH31039.03
SH31037.01EA 506 EUR
SH31037.01 SH31039.04 SH31037.08 SH31039.03
Cell culture medium, HyClone™ ActiPro
Zellkulturmedien
HyClone™ ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines.
  • Demonstrated high yields of recombinant proteins
  • Does not contain hypoxanthine and thymidine
  • Chemically defined ADCF formulation
  • Production medium for batch or fed-batch processes
  • Accompanied with detailed protocols for use to maximise output
HyClone™ ActiPro does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable