Cell culture medium, HyClone™ ActiPro
Lieferant: HyClone products (Cytiva)
SH31037.01
SH31039.04
SH31037.08
SH31039.03
SH31037.01EA
506
EUR
SH31037.01
SH31039.04
SH31037.08
SH31039.03
Cell culture medium, HyClone™ ActiPro
Zellkulturmedien
HyClone™ ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines.
- Demonstrated high yields of recombinant proteins
- Does not contain hypoxanthine and thymidine
- Chemically defined ADCF formulation
- Production medium for batch or fed-batch processes
- Accompanied with detailed protocols for use to maximise output
HyClone™ ActiPro does not contain any growth factors such as insulin, peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.