HyClone™ VaccineXpress Cell Culture Media
Lieferant: HyClone products (Cytiva)
SH31126.01
SH31126.01EA 242 EUR
SH31126.01
HyClone™ VaccineXpress Cell Culture Media
Zellkulturmedien
HyClone VaccineXpress cell culture medium is designed and developed for high‑density growth and maintenance of kidney‑derived cell lines (e.g., Vero cells) for viral vaccine development. VaccineXpress medium is well suited for use in multi-well plates and large‑scale manufacturing using microcarriers in WAVE Bioreactor or Xcellerex bioreactors.
  • Serum-free animal-derived component-free formulation
  • Supports direct or sequential adaptation, cell growth and virus propagation
  • Designed to support high peak cell density, viral infectivity, and productivity
  • For use with multiple viral applications (i.e., influenza, rotavirus, RSV, Dengue, Zika)
  • Designed for small- to large-scale cell culture applications with use of microcarriers
  • Available in liquid and powder formulations
HyClone VaccineXpress medium has been qualified to grow attachment‑dependent Vero cells with microcarriers for producing viral vaccines. The lack of both serum and animal‑derived components in the medium reduces variability and enhances scalability and ease of purifying recombinant proteins and viruses in bioprocess applications. VaccineXpress is formulated without L‑glutamine for extended shelf life.