Natriumchlorid-Lösung 1 M, in single-use containers, HyClone™ für Bioverfahrenstechnik

Lieferant: HyClone products (Cytiva)

SH31140.01 SH31140.02 SH31140.03
SH31140.01EA 616 EUR
SH31140.01 SH31140.02 SH31140.03
Natriumchlorid-Lösung 1 M, in single-use containers, HyClone™ für Bioverfahrenstechnik
Natriumchlorid

HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.


  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 µm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.


These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.

Formel: NaCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5

Bestellen

Spezifikation Testergebnisse

Storage Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
Endotoxin Concentration ≤ 1 EU/mL
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 Months
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR